Anti-IL32 Mouse Monoclonal Antibody [clone: AT2F9]
Catalog # 10720-996
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 32
- Antigen Symbol:IL32
- Clonality:Monoclonal
- Clone:AT2F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:9235
- Antigen Synonyms:TAIFd|IL-32|IL-32alpha|Interleukin-32|TAIFa|TAIFc|IL-32beta|IL-32gamma|NK4|TAIF|Natural killer cells protein 4|TAIFb|IL-32delta
- Amino Acid Number:1 to 131
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-131 of human IL32.
- Purification:Protein G purification
- Cat. No.:10720-996
- Supplier no.:MAB9712
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant IL32.
Recommended Dilutions: ELISA: 1:500; Western Blot: 1:500
Type: Primary
Antigen: IL32
Clonality: Monoclonal
Clone: AT2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human