Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-PFKFB1 Rabbit Polyclonal Antibody
Catalog # 10682-272
Supplier:  Abnova
undefined
Anti-PFKFB1 Rabbit Polyclonal Antibody
Catalog # 10682-272
Supplier:  Abnova
Supplier Number:  PAB4021

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
  • Antigen Symbol:
    PFKFB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5207
  • Antigen Synonyms:
    6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1|F6PK|6PF-2-K/Fru-2,6-P2ASE liver isozyme
  • Storage Buffer:
    PBS (0.09% sodium azide)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to internal region of human PFKFB1.
  • Purification:
    Protein G purification
  • Cat. No.:
    10682-272
  • Supplier no.:
    PAB4021

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of PFKFB1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: PFKFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human