To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Gamma
- Antigen Symbol:PIK3CG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5294
- Antigen Synonyms:PtdIns-3-kinase subunit p110|PI3-kinase p110 subunit gamma|PI3K|PI3Kgamma|Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform|PIK3CG|p120-PI3K
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PIK3CG.
- Purification:Protein G purification
- Cat. No.:10679-650
- Supplier no.:PAB3218
Rabbit polyclonal antibody raised against synthetic peptide of PIK3CG.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100
Type: Primary
Antigen: PIK3CG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human