Anti-GRIN3B Goat Polyclonal Antibody
Catalog # 10716-518
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor, Ionotropic, N-methyl-D-Aspartate 3B
- Antigen Symbol:GRIN3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Liquid
- Gene ID:116444
- Antigen Synonyms:GluN3B|NR3B
- Storage Buffer:0.5 mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
- Sequence:C-EAPPHSGRPGSQ
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at C-terminus of human GRIN3B.
- Purification:Antigen affinity purification
- Cat. No.:10716-518
- Supplier no.:PAB18635
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of GRIN3B.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.5-1.5 ?g/ml
Type: Primary
Antigen: GRIN3B
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope: C-terminal
Host:
Isotype: Goat
Reactivity: Human