Anti-LCMT2 Rabbit Polyclonal Antibody
Catalog # 10723-388
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine carboxyl methyltransferase 2
- Antigen Symbol:LCMT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:9836
- Antigen Synonyms:Leucine carboxyl methyltransferase 2|KIAA0547|PPM2|TYW4|tRNA yW-synthesizing protein 4|tRNA wybutosine-synthesizing protein 4
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids near N-terminus of human LCMT2.
- Purification:Peptide affinity purification
- Cat. No.:10723-388
- Supplier no.:PAB19442
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of LCMT2.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 5 ?g/ml
Type: Primary
Antigen: LCMT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat