Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # 10733-816
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta
- Antigen Symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5564
- Antigen Synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Storage Buffer:PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human PRKAB1.
- Purification:Affinity purification
- Cat. No.:10733-816
- Supplier no.:PAB18144
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of PRKAB1.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:20000
Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat