Anti-C2ORF40 Rabbit Polyclonal Antibody
Catalog # 10704-192
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Chromosome 2 open reading frame 40
- Antigen Symbol:C2ORF40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:84417
- Antigen Synonyms:ECRG4
- Storage Buffer:PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human C2orf40.
- Cat. No.:10704-192
- Supplier no.:PAB17451
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of C2orf40.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:40000
Type: Primary
Antigen: C2ORF40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse