Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # 10733-818
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta
- Antigen Symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:5564
- Antigen Synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Storage Buffer:0.02% sodium azide
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at N-terminus of human PRKAB1.
- Cat. No.:10733-818
- Supplier no.:PAB19838
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of PRKAB1.
Recommended Dilutions: ELISA: 1:80000; Western Blot: 1:200-1000
Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human