Anti-HAPLN4 Mouse Monoclonal Antibody [clone: 1C1]
Catalog # 10588-310
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:hyaluronan and proteoglycan link protein 4
- Antigen Symbol:HAPLN4
- Clonality:Monoclonal
- Clone:1C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:404037
- Antigen Synonyms:BRAL2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10588-310
- Supplier no.:H00404037-M02
Specifications
About this item
Type: Primary
Antigen: HAPLN4 (hyaluronan and proteoglycan link protein 4)
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human