Anti-TSEN54 Mouse Polyclonal Antibody
Catalog # 10575-002
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:TSEN54 tRNA splicing endonuclease Subunit
- Antigen Symbol:TSEN54
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:283989
- Antigen Synonyms:SEN54L|sen54|PCH4|PCH2A|PCH5
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10575-002
- Supplier no.:H00283989-A01
Specifications
About this item
Type: Primary
Antigen: TSEN54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human