Anti-RNF166 Mouse Polyclonal Antibody
Catalog # 10570-048
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ring Finger Protein 166
- Antigen Symbol:RNF166
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:115992
- Antigen Synonyms:Zfp313l|Zinc finger protein 313.|RNF 166|RNF166|RING finger protein 166|RN166_HUMAN|Zinc finger protein 228
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10570-048
- Supplier no.:H00115992-B01P
Specifications
About this item
Type: Primary
Antigen: RNF166
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human