Anti-ZNF436 Mouse Polyclonal Antibody
Catalog # 10574-594
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 436
- Antigen Symbol:ZNF436
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:80818
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10574-594
- Supplier no.:H00080818-B01P
Specifications
About this item
Type: Primary
Antigen: ZNF436
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human