Anti-MBD3 Rabbit Polyclonal Antibody
Catalog # 10592-986
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl-CpG-binding Domain protein 3
- Antigen Symbol:MBD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Gene ID:53615
- Antigen Synonyms:methyl-CpG-binding domain protein 3|CpG-binding domain protein 3|CpG-binding domain 3
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10592-986
- Supplier no.:H00053615-D01
Specifications
About this item
Type: Primary
Antigen: MBD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human