Anti-PRKAG2 Mouse Monoclonal Antibody [clone: 3C4]
Catalog # 10598-068
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 2
- Antigen Symbol:PRKAG2
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:51422
- Antigen Synonyms:CMH6|AAKG|AAKG2|WPWS|H91620p
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-068
- Supplier no.:H00051422-M01
Specifications
About this item
Type: Primary
Antigen: PRKAG2
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human