Anti-GPKOW Mouse Polyclonal Antibody
Catalog # 10557-736
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:G patch domain and KOW motifs
- Antigen Symbol:GPKOW
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:27238
- Antigen Synonyms:Spp2|T54|GPATC5|GPATCH5
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10557-736
- Supplier no.:H00027238-B01
Specifications
About this item
Type: Primary
Antigen: GPKOW (G patch domain and KOW motifs)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human