Anti-RABGAP1 Rabbit Polyclonal Antibody
Catalog # 10599-006
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:RAB GTPase activating Protein 1
- Antigen Symbol:RABGAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Gene ID:23637
- Antigen Synonyms:HSPC094|Rab6 GTPase activating protein GAPCenA|RBGP1_HUMAN.|Rab GTPase activating protein 1|GAP and centrosome associated protein
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10599-006
- Supplier no.:H00023637-D01
Specifications
About this item
Type: Primary
Antigen: RABGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human