Anti-MSRB2 Mouse Monoclonal Antibody [clone: 3F12]
Catalog # 10593-918
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methionine Sulfoxide Reductase B2
- Antigen Symbol:MSRB2
- Clonality:Monoclonal
- Clone:3F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:22921
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10593-918
- Supplier no.:H00022921-M03
Specifications
About this item
Type: Primary
Antigen: MSRB2
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human