Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB Rabbit Polyclonal Antibody
Anti-ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB Rabbit Polyclonal Antibody
Catalog # 10576-004
Supplier:  Abnova
undefined
Anti-ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB Rabbit Polyclonal Antibody
Catalog # 10576-004
Supplier:  Abnova
Supplier Number:  H00010152-D01P

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Abl-Interactor 2
  • Antigen Symbol:
    ABI2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Gene ID:
    10152
  • Antigen Synonyms:
    argBPIA|ABI2B|AIP-1|ABI-2|argBPIB|AblBP3|SSH3BP2|argBP1
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10576-004
  • Supplier no.:
    H00010152-D01P

Specifications

About this item

Type: Primary
Antigen: ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human