Anti-ELL Mouse Polyclonal Antibody
10555-674
: Abnova
- Antibody Type:Primary
- Antigen Name:elongation factor RNA polymerase II
- Antigen Symbol:ELL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:8178
- Antigen Synonyms:PPP1R68|MEN|C19orf17|ELL1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10555-674
- Supplier no.:H00008178-A01
Type: Primary
Antigen: ELL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human