Anti-UCHL1 Mouse Monoclonal Antibody [clone: 1B8-4D2]
Catalog # 10605-308
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen Symbol:UCHL1
- Clonality:Monoclonal
- Clone:1B8-4D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:7345
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10605-308
- Supplier no.:H00007345-M01
Specifications
About this item
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 1B8-4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human