Anti-VPS72 Mouse Monoclonal Antibody [clone: 2G6]
10603-168
: Abnova
- Antibody Type:Primary
- Antigen Name:Vacuolar Protein Sorting 72
- Antigen Symbol:VPS72
- Clonality:Monoclonal
- Clone:2G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:6944
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10603-168
- Supplier no.:H00006944-M01
Type: Primary
Antigen: VPS72
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human