Anti-PRKAB2 Mouse Monoclonal Antibody [clone: 2G9]
10598-050
: Abnova
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 2
- Antigen Symbol:PRKAB2
- Clonality:Monoclonal
- Clone:2G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5565
- Antigen Synonyms:BB124140|5730553K21Rik|AW049591
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-050
- Supplier no.:H00005565-M01
Type: Primary
Antigen: PRKAB2
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human