Anti-PRKAB2 Mouse Polyclonal Antibody
10568-392
: Abnova
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 2
- Antigen Symbol:PRKAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:5565
- Antigen Synonyms:BB124140|5730553K21Rik|AW049591
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10568-392
- Supplier no.:H00005565-A01
Type: Primary
Antigen: PRKAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human