Anti-PRKAB1 Mouse Monoclonal Antibody [clone: 1D3]
10598-046
: Abnova
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta
- Antigen Symbol:PRKAB1
- Clonality:Monoclonal
- Clone:1D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5564
- Antigen Synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-046
- Supplier no.:H00005564-M07
Type: Primary
Antigen: PRKAB1
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse