Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # 10598-048
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta
- Antigen Symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5564
- Antigen Synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-048
- Supplier no.:H00005564-D01P
Specifications
About this item
Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human