Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # 10598-040
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen Symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5563
- Antigen Synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-040
- Supplier no.:H00005563-D01P
Specifications
About this item
Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human