Anti-PRKAA1 Mouse Polyclonal Antibody
Catalog # 10568-388
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:5562
- Antigen Synonyms:catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10568-388
- Supplier no.:H00005562-B01P
Specifications
About this item
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human