Anti-PDE3A Mouse Monoclonal Antibody [clone: 2D7]
Catalog # 10596-424
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphodiesterase 3A, cGMP-stimulated
- Antigen Symbol:PDE3A
- Clonality:Monoclonal
- Clone:2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5139
- Antigen Synonyms:CGI-PDE-A|HTNB|CGI-PDE|CGI-PDE A
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10596-424
- Supplier no.:H00005139-M03
Specifications
About this item
Type: Primary
Antigen: PDE3A
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human