Anti-MECP2 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # 10593-102
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl CPG Binding Protein 2
- Antigen Symbol:MECP2
- Clonality:Monoclonal
- Clone:4B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:4204
- Antigen Synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10593-102
- Supplier no.:H00004204-M01
Specifications
About this item
Type: Primary
Antigen: MECP2
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat