Anti-MECP2 Mouse Polyclonal Antibody
Catalog # 10563-986
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl CPG Binding Protein 2
- Antigen Symbol:MECP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:4204
- Antigen Synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10563-986
- Supplier no.:H00004204-B01P
Specifications
About this item
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human