Anti-MBD1 Mouse Monoclonal Antibody [clone: 2B10]
Catalog # 10592-978
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl-CpG Binding Domain Protein 1
- Antigen Symbol:MBD1
- Clonality:Monoclonal
- Clone:2B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:4152
- Antigen Synonyms:CXXC3|PCM1|RFT
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10592-978
- Supplier no.:H00004152-M05
Specifications
About this item
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human