Anti-GRIN2C Mouse Polyclonal Antibody
10557-896
: Abnova
- Antibody Type:Primary
- Antigen Name:Glutamate Ionotropic Receptor NMDA Type Subunit 2C
- Antigen Symbol:GRIN2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:2905
- Antigen Synonyms:NMDAR2C|GluN2C|NR2C
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10557-896
- Supplier no.:H00002905-B01P
Type: Primary
Antigen: GRIN2C (glutamate receptor, ionotropic, N-methyl D-aspartate 2C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human