Anti-GRIN2B Mouse Monoclonal Antibody [clone: 2G5]
Catalog # 10587-918
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Ionotropic Receptor NMDA Type Subunit 2B
- Antigen Symbol:GRIN2B
- Clonality:Monoclonal
- Clone:2G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:2904
- Antigen Synonyms:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10587-918
- Supplier no.:H00002904-M01
Specifications
About this item
Type: Primary
Antigen: GRIN2B (glutamate receptor, ionotropic, N-methyl D-aspartate 2B)
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human