Anti-ACSL3 Mouse Polyclonal Antibody
Catalog # 10510-232
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Acyl-CoA Synthetase Long-chain Family Member 3
- Antigen Symbol:ACSL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:2181
- Antigen Synonyms:ACSL3|ACS3|LACS3|Long-chain acyl-CoA synthetase 3
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10510-232
- Supplier no.:H00002181-A01
Specifications
About this item
Type: Primary
Antigen: ACSL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human