Anti-SLC26A2 Mouse Monoclonal Antibody [clone: 3F6]
Catalog # 10601-032
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 26 Member A2
- Antigen Symbol:SLC26A2
- Clonality:Monoclonal
- Clone:3F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:1836
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10601-032
- Supplier no.:H00001836-M04
Specifications
About this item
Type: Primary
Antigen: SLC26A2
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human