Anti-CYP4B1 Mouse Polyclonal Antibody
Catalog # 10554-662
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome P450, Family 4, Subfamily B, Polypeptide 1
- Antigen Symbol:CYP4B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:1580
- Antigen Synonyms:Cytochrome P450 4B1|CYPIVB1|Cytochrome P450-HP|CYP4B1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10554-662
- Supplier no.:H00001580-B01
Specifications
About this item
Type: Primary
Antigen: CYP4B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: