Anti-ASNS Rabbit Polyclonal Antibody
Catalog # 10581-024
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Asparagine Synthetase (Glutamine-Hydrolyzing)
- Antigen Symbol:ASNS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Gene ID:440
- Antigen Synonyms:ASNSD|TS11
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10581-024
- Supplier no.:H00000440-D01
Specifications
About this item
Type: Primary
Antigen: ASNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human