Anti-TRIM23 Mouse Monoclonal Antibody [clone: 2G4]
10577-196
: Abnova
- Antibody Type:Primary
- Antigen Name:Tripartite Motif Containing 23
- Antigen Symbol:TRIM23
- Clonality:Monoclonal
- Clone:2G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:373
- Antigen Synonyms:RNF46|ARFD1|ARD1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10577-196
- Supplier no.:H00000373-M12
Type: Primary
Antigen: TRIM23
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human