Anti-AMPD2 Mouse Monoclonal Antibody [clone: 6A8]
10623-842
: Abnova
- Antibody Type:Primary
- Antigen Name:AMP deaminase 2
- Antigen Symbol:AMPD2
- Clonality:Monoclonal
- Clone:6A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Cross Adsorption:No
- Gene ID:271
- Antigen Synonyms:PCH9|SPG63
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10623-842
- Supplier no.:H00000271-M09A
Type: Primary
Antigen: AMPD2
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human