Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 2F5]
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # 10551-322
Supplier:  Abnova
undefined
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # 10551-322
Supplier:  Abnova
Supplier Number:  H00000271-M01

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AMP deaminase 2
  • Antigen Symbol:
    AMPD2
  • Clonality:
    Monoclonal
  • Clone:
    2F5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Cross Adsorption:
    No
  • Gene ID:
    271
  • Antigen Synonyms:
    PCH9|SPG63
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10551-322
  • Supplier no.:
    H00000271-M01

Specifications

About this item

Type: Primary
Antigen: AMPD2
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat