Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AMPD2 Rabbit Polyclonal Antibody
Anti-AMPD2 Rabbit Polyclonal Antibody
Catalog # 10576-806
Supplier:  Abnova
undefined
Anti-AMPD2 Rabbit Polyclonal Antibody
Catalog # 10576-806
Supplier:  Abnova
Supplier Number:  H00000271-D01

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AMP deaminase 2
  • Antigen Symbol:
    AMPD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Gene ID:
    271
  • Antigen Synonyms:
    PCH9|SPG63
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10576-806
  • Supplier no.:
    H00000271-D01

Specifications

About this item

Type: Primary
Antigen: AMPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human