Orders LinkContactUsLinkComponent
Attached Top Distribution Containers, Quantum Storage Systems
27 5/16×16 9/16×12 1/2" (E×terior), 25 1/8×15 3/8×12" (Interior)
  10752-914
 :  QDC2717-12
Attached Top Distribution Containers, Quantum Storage Systems
  10752-914
Attached Top Distribution Containers, Quantum Storage Systems
Attached Top Distribution Containers, Quantum Storage Systems
27 5/16×16 9/16×12 1/2" (E×terior), 25 1/8×15 3/8×12" (Interior)
$74.93
Case of 1 Each
Strong, reliable and reusable totes are ideal for storage, distribution and order picking.
  • Stack for maximized storage and nest when empty
  • Textured, non-slip bottom ensures smooth conveying
  • Ergonomically designed handles allow for easy lifting
  • Recessed lids ensure maximum load stability
  • Padlock eye provides security option allowing lids to be fastened and/or locked
  • Lids close securely to protect from dust or damage
  • Molded from high density, FDA approved polyethlene
Tops are permanently attached with hinges that interlock for stacking and security. Gray color.
  • Dimensions:
    27 5/16×16 9/16×12 1/2" (E×terior), 25 1/8×15 3/8×12" (Interior)