Stack and Nest Totes, Quantum Storage Systems
Supplier: QUANTUM STORAGE SYSTEMS
Certificates
About this item
Heavy-duty and dependable stack and nest totes are reusable container for storing and transporting your valuable items in any application.
- Available in red, blue, or gray
- Available in nine sizes
- Unique ability to stack or they can be turned 180o to nest when empty saving valuable space without using a lid
- Lids available
Injection molded from high-density polypropylene that complies with the FDA and USDA regulations.
Frequently Bought Together