Human Recombinant IL-4 (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤0.25 ng/ml, Bioactivity ≥4.0×10⁶ units/mg
- Gene ID:P05112
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:BCGF|B-cell stimulatory factor-1|BCDF|BSF-1
- Protein/Peptide Name:IL-4
- Purity:≥95%
- Molecular Weight:15.1 kDa
- Sequence:MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Related Information
Frequently Bought Together