MetriCide™ 28 High-Level Disinfectant, Metrex
Supplier: Metrex
Certificates
About this item
MetriCide™ 28 is a 2.5% glutaraldehyde solution which can be used for the sterilization and high-level disinfection of various medical devices for up to 28 days.
- Contains surfactants
- When activated, solution attains an alkaline pH between 7.5 and 8.5
- Activation required
- Recommended for immersible and heat-sensitive equipment and instruments
It is intended for use in a tray system with a variety of non-critical, semi-critical, and critical devices. This includes anesthesia equipment, respiratory therapy equipment, metallic equipment or instruments, rubber objects, plastic objects, and thermometers.