Order Entry
Puerto Rico
ContactUsLinkComponent
RBC Lysis Buffer (10X), Biolegend
RBC Lysis Buffer (10X), Biolegend
Catalog # 420301-BL
Supplier:  Biolegend
CAS Number:  
RBC Lysis Buffer (10X), Biolegend
Catalog # 420301-BL
Supplier:  Biolegend
Supplier Number:  420301
CAS Number:  

Specifications

  • Description:
    RBC Lysis Buffer (10X)
  • Size:
    100 mL
  • Storage Temperature:
    Store at 2 to 8°C
  • Cat. No.:
    420301-BL
  • Supplier no.:
    420301
  • Reactivity:
    Mouse, Human

Specifications

About this item

Red Blood Cell (RBC) Lysis Buffer has been designed, formulated, and tested to ensure optimal lysis of RBCs in single cell suspensions with minimal effects on leukocytes.

  • Tested to ensure optimal lysis of RBCs in single cell suspensions
  • Minimal effects on leukocytes

RBC Lysis Buffer is supplied as a 10X solution containing ammonium chloride, potassium carbonate, and EDTA, and should be diluted in deionized water prior to use. Nucleated RBCs are not effectively lysed with ammonium chloride.