Quantofix EDTA Test Strips, CTL Scientific Supply
Catalog # 10217-566
Supplier: Macherey-Nagel
CAS Number:
Specifications
- Description:EDTA Test Strips
- Storage:RT
- Cat. No.:10217-566
- Range:0-100-200-300-400 mg/L
- Supplier no.:91335
Specifications
About this item
Quantofix EDTA test strips is for the rapid and reliable determination of EDTA, NTA and other complexing agents in solutions
Quantofix EDTA has a gradation of scale: 0-100-200-300-400 mg/L EDTA.
The easy dip and read procedure provides a reliable result within 15 seconds.
Ordering information: Packaged as a box of 100.