Quantofix® Zinc Test Kit, CTL Scientific Supply
Catalog # 10217-564
Supplier: Macherey-Nagel
CAS Number:
Specifications
- Description:Zinc Test Kit
- Storage:RT
- Cat. No.:10217-564
- Range:0-2-5-10-25-50-100 mg/L
- Supplier no.:91310
Specifications
About this item
Quantofix Zinc test strips and reagent for the semi-quantitative determination of zinc (Zn2+) in solutions
Quantofix Zinc has a gradation of scale: 0-2-5-10-25-50-100 mg/L Zn2+ orange to red.
The following ions only interfere in larger concentrations: > 1000 mg/L Ag+, Al3+, Bi3+, Ca2+, Cd2+, Co2+, Fe2+, Fe3+, Mn2+, NH4+, Pb2+, Sn2+, Sn4+, Cl-, CrO42-, NO2 -, NO3 -, PO43-, SCN-, SO32-, SO42-, acetate, ascorbate, citrate, oxalate, tartrate. The following ions will cause low mg/L Ni2+. The presence of Cu2+, >10 mg/L Hg+ /Hg2+ (precipitation with iron or cadmium powder in weakly acidic solution) and >50 mg/L MnO4- (destroy with hydroxylammonium chloride in acidic solution) cause a brown coloration of the test field.
Ordering information: Packaged as a box of 100, strips and reagent.