ContactUsLinkComponent
Quantofix Tin Test Strips, CTL Scientific Supply
Quantofix Tin Test Strips, CTL Scientific Supply
  10217-562
 :  
undefined
Quantofix Tin Test Strips, CTL Scientific Supply
  10217-562
 :  91309
 :  
  • Description:
    Tin Test Strips
  • Storage:
    RT
  • Cat. No.:
    10217-562
  • Range:
    0-10-25-50-100250-500 mg/L
  • Supplier no.:
    91309
Quantofix Tin Test strips for the semi-quantitative determination of tin (ll)-ions in solutions.
Quantofix Tin has a gradation of scale: 0-10-25-50-100250-500 mg/L Sn2+ white to dark blue.
 : Test procedure is intricate; product not recommended for use by non-chemist. Request test procedure prior to ordering. Contact your Avantor representative for more information. Packaged as a box of 100.