Order Entry
Puerto Rico
ContactUsLinkComponent
Quantofix Glutaraldehyde Test Strips, CTL Scientific Supply
Quantofix Glutaraldehyde Test Strips, CTL Scientific Supply
Catalog # 10217-550
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Quantofix Glutaraldehyde Test Strips, CTL Scientific Supply
Catalog # 10217-550
Supplier:  Macherey-Nagel
Supplier Number:  91343
CAS Number:  

Specifications

  • Description:
    Glutaraldehyde Test Strips
  • Storage:
    RT
  • Cat. No.:
    10217-550
  • Range:
    0-0.5-1-1.5-2-2.5 mg/L
  • Supplier no.:
    91343

Specifications

About this item

Quantofix Glutaraldehyde test strips for the semi-quantitative determination of glutaraldehyde.

Quantofix Glutaraldehyde test strips has a gradation of scale: 0-0.5-1-1.5-2-2.5 mg/L.

Ordering information: Packaged as a box of 100.